Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacng7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079281.1 | NP_001072749.1 | 2 - 3 | 598 | 83 | |
GenBank mRNA | BC125751.1 | - | 2 - 3 | 598 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |