Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079346.2 | NP_001072814.2 | 3 - 4 | 582 | 100 | |
GenBank mRNA | BC121592.1 | - | 3 - 4 | 558 | 100 | |
BC167921.1 | - | 3 - 4 | 523 | 100 | ||
CN089555.1 | - | 3 - 4 | 582 | 100 | ||
CR848177.2 | - | 3 - 4 | 552 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |