Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fezf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016305.3 | NP_001016305.1 | 3 - 4 | 1182 | 100 | |
XM_012961082.2 | XP_012816536.1 | 2 - 3 | 1203 | 100 | ||
GenBank mRNA | BC136188.1 | - | 3 - 4 | 1182 | 100 | |
CR761501.2 | - | 2 - 3 | 935 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |