Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr2c2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016306.3 | NP_001016306.1 | 3 - 4 | 323 | 87 | |
GenBank mRNA | BC129029.1 | - | 3 - 4 | 323 | 87 | |
CR761499.2 | - | 3 - 4 | 373 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |