Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b3glct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079083.1 | NP_001072551.1 | 3 - 4 | 203 | 102 | |
XM_012957278.2 | XP_012812732.1 | 2 - 3 | 296 | 102 | ||
GenBank mRNA | BC122064.1 | - | 3 - 4 | 203 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |