Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b3glct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079083.1 | NP_001072551.1 | 9 - 10 | 834 | 74 | |
XM_012957278.2 | XP_012812732.1 | 8 - 9 | 927 | 74 | ||
XM_018091349.1 | XP_017946838.1 | 7 - 8 | 658 | 74 | ||
GenBank mRNA | BC122064.1 | - | 9 - 10 | 834 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |