Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coq10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079105.1 | NP_001072573.1 | 2 - 3 | 399 | 90 | |
GenBank mRNA | BC122920.1 | - | 2 - 3 | 399 | 90 | |
CT010563.2 | - | 2 - 3 | 378 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |