Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ostm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079114.1 | NP_001072582.1 | 5 - 6 | 1155 | 60 | |
GenBank mRNA | BC122941.1 | - | 5 - 6 | 1155 | 60 | |
CT485681.1 | - | 5 - 6 | 1141 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |