Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab21l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079329.1 | NP_001072797.1 | 1 | 1048 | 99 | |
GenBank mRNA | BC121357.1 | - | 1 | 1048 | 99 | |
BC160505.1 | - | 1 | 1008 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |