Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079424.1 | NP_001072892.1 | 3 - 4 | 651 | 69 | |
GenBank mRNA | BC125782.1 | - | 3 - 4 | 651 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |