Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083354.2 | NP_001076823.1 | 11 - 12 | 1846 | 63 | |
GenBank mRNA | BC135750.1 | - | 11 - 12 | 1846 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |