Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083354.2 | NP_001076823.1 | 5 - 6 | 983 | 107 | |
XM_012959159.2 | XP_012814613.1 | 5 - 6 | 1512 | 107 | ||
GenBank mRNA | BC135750.1 | - | 5 - 6 | 983 | 107 | |
CR761177.2 | - | 5 - 6 | 930 | 107 | ||
CT485733.1 | - | 5 - 6 | 935 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |