Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: etv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097229.1 | NP_001090698.1 | 3 - 4 | 330 | 76 | |
XM_012967807.2 | XP_012823261.1 | 3 - 4 | 468 | 76 | ||
GenBank mRNA | BC128620.1 | - | 3 - 4 | 330 | 76 | |
CT030495.1 | - | 2 - 3 | 308 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |