Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097231.1 | NP_001090700.1 | 1 - 2 | 487 | 92 | |
XM_018095197.1 | XP_017950686.1 | 1 - 2 | 789 | 92 | ||
GenBank mRNA | BC128622.1 | - | 1 - 2 | 487 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |