Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nop16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102748.1 | NP_001096218.1 | 1 - 2 | 138 | 66 | |
GenBank mRNA | BC068216.1 | - | 1 - 2 | 138 | 66 | |
CR848327.2 | - | 1 - 2 | 132 | 66 | ||
CU025085.1 | - | 1 - 2 | 138 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |