Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: blvrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102799.1 | NP_001096269.1 | 2 - 3 | 238 | 99 | |
XM_012967878.1 | XP_012823332.1 | 2 - 3 | 326 | 99 | ||
GenBank mRNA | BC135495.1 | - | 2 - 3 | 238 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |