Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rilpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102850.1 | NP_001096320.1 | 5 - 6 | 1152 | 102 | |
GenBank mRNA | BC127552.1 | - | 5 - 6 | 1152 | 102 | |
BC135712.1 | - | 5 - 6 | 1136 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |