Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102851.1 | NP_001096321.1 | 9 - 10 | 1419 | 80 | |
XM_012958422.2 | XP_012813876.1 | 9 - 10 | 1712 | 80 | ||
GenBank mRNA | BC135716.1 | - | 9 - 10 | 1419 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |