Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102859.1 | NP_001096329.1 | 3 - 4 | 451 | 74 | |
GenBank mRNA | BC135747.1 | - | 3 - 4 | 451 | 74 | |
CR848288.2 | - | 3 - 4 | 398 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |