Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plppr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102900.1 | NP_001096370.1 | 3 - 4 | 208 | 104 | |
XM_012964809.2 | XP_012820263.1 | 2 - 3 | 454 | 104 | ||
GenBank mRNA | BC135902.1 | - | 3 - 4 | 208 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |