Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: samd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102935.1 | NP_001096405.1 | 2 - 3 | 218 | 78 | |
XM_012965901.2 | XP_012821355.1 | 7 - 8 | 1989 | 78 | ||
XM_012965902.2 | XP_012821356.1 | 7 - 8 | 1938 | 78 | ||
XM_012965903.2 | XP_012821357.1 | 6 - 7 | 1890 | 78 | ||
XM_012965904.2 | XP_012821358.1 | 7 - 8 | 2002 | 78 | ||
XM_012965906.2 | XP_012821360.1 | 7 - 8 | 748 | 78 | ||
XM_012965908.2 | XP_012821362.1 | 7 - 8 | 697 | 78 | ||
XM_012965909.2 | XP_012821363.1 | 7 - 8 | 678 | 78 | ||
XM_012965910.2 | XP_012821364.1 | 7 - 8 | 685 | 78 | ||
XM_012965911.2 | XP_012821365.1 | 7 - 8 | 678 | 78 | ||
XM_012965912.2 | XP_012821366.1 | 7 - 8 | 618 | 78 | ||
XM_012965913.2 | XP_012821367.1 | 7 - 8 | 628 | 78 | ||
XM_012965914.2 | XP_012821368.1 | 6 - 7 | 577 | 78 | ||
GenBank mRNA | BC136045.1 | - | 2 - 3 | 218 | 78 | |
CR942606.2 | - | 2 - 3 | 233 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |