Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam210a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103011.1 | NP_001096481.1 | 3 - 4 | 599 | 84 | |
XM_012964207.2 | XP_012819661.1 | 3 - 4 | 699 | 84 | ||
XM_012964208.2 | XP_012819662.1 | 2 - 3 | 555 | 84 | ||
GenBank mRNA | BC136245.1 | - | 3 - 4 | 599 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |