Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nubp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103064.1 | NP_001096534.1 | 2 - 3 | 147 | 59 | |
XM_012969994.2 | XP_012825448.1 | 2 - 3 | 223 | 59 | ||
XM_018097028.1 | XP_017952517.1 | 2 - 3 | 525 | 59 | ||
GenBank mRNA | BC135883.1 | - | 2 - 3 | 147 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |