Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl6ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016131.5 | NP_001016131.1 | 5 - 6 | 596 | 79 | |
GenBank mRNA | BC118695.2 | - | 5 - 6 | 597 | 79 | |
CR762046.2 | - | 5 - 6 | 508 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |