Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016782.4 | NP_001016782.1 | 3 - 4 | 147 | 77 | |
XM_012968442.2 | XP_012823896.1 | 3 - 4 | 222 | 77 | ||
GenBank mRNA | AL969102.2 | - | 138 | 77 | ||
BC157798.1 | - | 3 - 4 | 147 | 77 | ||
CX484399.2 | - | 3 - 4 | 147 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |