Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103071.1 | NP_001096541.1 | 7 - 8 | 572 | 66 | |
XM_012971507.2 | XP_012826961.1 | 7 - 8 | 583 | 66 | ||
GenBank mRNA | BC135970.1 | - | 7 - 8 | 572 | 66 | |
CT025394.2 | - | 7 - 8 | 584 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |