Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103072.1 | NP_001096542.1 | 2 - 3 | 186 | 69 | |
XM_012956320.2 | XP_012811774.1 | 2 - 3 | 297 | 69 | ||
GenBank mRNA | BC136023.1 | - | 2 - 3 | 186 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |