Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105284.1 | NP_001098754.1 | 8 - 9 | 683 | 80 | |
GenBank mRNA | BC135931.1 | - | 8 - 9 | 670 | 80 | |
BC152057.1 | - | 8 - 9 | 683 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |