Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab11fip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112943.1 | NP_001106414.1 | 3 - 4 | 1163 | 62 | |
XM_012953956.2 | XP_012809410.1 | 4 - 5 | 1390 | 62 | ||
XM_012953957.2 | XP_012809411.1 | 4 - 5 | 1425 | 62 | ||
XM_012953958.2 | XP_012809412.1 | 5 - 6 | 1507 | 62 | ||
XM_012953959.2 | XP_012809413.1 | 5 - 6 | 1542 | 62 | ||
GenBank mRNA | BC118910.1 | - | 3 - 4 | 1164 | 62 | |
BC153348.1 | - | 3 - 4 | 1163 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |