Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113678.1 | NP_001107150.1 | 3 - 4 | 287 | 87 | |
GenBank mRNA | AM041994.1 | - | 3 - 4 | 287 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Xenopus tropicalis Probe spans exons |