Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113681.1 | NP_001107153.1 | 2 - 3 | 234 | 67 | |
XM_012956377.2 | XP_012811831.1 | 3 - 4 | 525 | 67 | ||
XM_012956378.2 | XP_012811832.1 | 2 - 3 | 478 | 67 | ||
GenBank mRNA | BC157232.1 | - | 2 - 3 | 234 | 67 | |
BC170913.1 | - | 2 - 3 | 177 | 67 | ||
BC170915.1 | - | 2 - 3 | 177 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |