Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mall | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113808.1 | NP_001107280.1 | 1 - 2 | 165 | 99 | |
GenBank mRNA | BC087766.1 | - | 1 - 2 | 151 | 99 | |
BC122087.1 | - | 1 - 2 | 145 | 99 | ||
BC154915.1 | - | 1 - 2 | 165 | 99 | ||
BC169149.1 | - | 1 - 2 | 134 | 99 | ||
BC171228.1 | - | 1 - 2 | 151 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |