Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113890.1 | NP_001107362.1 | 6 - 7 | 762 | 94 | |
XM_012960563.2 | XP_012816017.1 | 7 - 8 | 1308 | 94 | ||
XM_012960564.2 | XP_012816018.1 | 7 - 8 | 1356 | 94 | ||
XM_012960565.1 | XP_012816019.1 | 7 - 8 | 829 | 94 | ||
GenBank mRNA | BC082746.1 | - | 7 - 8 | 831 | 94 | |
BC157213.1 | - | 6 - 7 | 762 | 94 | ||
BC171074.1 | - | 6 - 7 | 655 | 94 | ||
BC171076.1 | - | 6 - 7 | 655 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |