Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113951.1 | NP_001107423.1 | 3 - 4 | 315 | 106 | |
XM_012971521.2 | XP_012826975.1 | 3 - 4 | 406 | 106 | ||
GenBank mRNA | BC157553.1 | - | 3 - 4 | 315 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |