Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114051.1 | NP_001107523.1 | 12 - 13 | 1128 | 89 | |
GenBank mRNA | BC157696.1 | - | 12 - 13 | 1128 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |