Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126727.1 | NP_001120199.1 | 2 - 3 | 244 | 88 | |
XM_012968002.2 | XP_012823456.1 | 2 - 3 | 326 | 88 | ||
XM_012968003.2 | XP_012823457.1 | 2 - 3 | 669 | 88 | ||
GenBank mRNA | BC159320.1 | - | 2 - 3 | 244 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |