Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126748.1 | NP_001120220.1 | 3 - 4 | 426 | 62 | |
GenBank mRNA | BC159399.1 | - | 3 - 4 | 426 | 62 |
Gene Symbol: LOC101732964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004920549.2 | XP_004920606.2 | 1 - 2 | 198 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |