Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126957.1 | NP_001120429.1 | 7 - 8 | 727 | 97 | |
XM_012966087.2 | XP_012821541.1 | 7 - 8 | 805 | 97 | ||
XM_012966088.2 | XP_012821542.1 | 7 - 8 | 806 | 97 | ||
XM_012966089.2 | XP_012821543.1 | 7 - 8 | 802 | 97 | ||
GenBank mRNA | BC161184.1 | - | 7 - 8 | 727 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |