Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127012.1 | NP_001120484.1 | 3 - 4 | 1097 | 75 | |
XM_012968029.2 | XP_012823483.1 | 4 - 5 | 796 | 75 | ||
XM_012968031.2 | XP_012823485.1 | 4 - 5 | 808 | 75 | ||
GenBank mRNA | BC161354.1 | - | 3 - 4 | 1097 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |