Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127056.1 | NP_001120528.1 | 3 - 4 | 352 | 63 | |
GenBank mRNA | CR762124.2 | - | 3 - 4 | 352 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis Probe spans exons |