Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127120.1 | NP_001120592.1 | 5 - 6 | 534 | 100 | |
XM_012968046.1 | XP_012823500.1 | 5 - 6 | 508 | 100 | ||
GenBank mRNA | BC161613.1 | - | 5 - 6 | 534 | 100 | |
BC167877.1 | - | 5 - 6 | 508 | 100 | ||
CU025132.1 | - | 5 - 6 | 511 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |