Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126752.1 | NP_001120224.1 | 3 - 4 | 1504 | 74 | |
GenBank mRNA | BC160387.1 | - | 3 - 4 | 1504 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |