Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126926.1 | NP_001120398.1 | 8 - 9 | 1858 | 99 | |
XM_018092927.1 | XP_017948416.1 | 8 - 9 | 1852 | 99 | ||
XM_018092928.1 | XP_017948417.1 | 8 - 9 | 1967 | 99 | ||
GenBank mRNA | BC161108.1 | - | 8 - 9 | 1858 | 99 | |
CR855483.2 | - | 4 - 5 | 680 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |