Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126935.1 | NP_001120407.1 | 2 - 3 | 274 | 81 | |
XM_012962732.2 | XP_012818186.1 | 2 - 3 | 354 | 81 | ||
XM_012962733.2 | XP_012818187.1 | 2 - 3 | 420 | 81 | ||
XM_012962734.2 | XP_012818188.1 | 2 - 3 | 318 | 81 | ||
XM_012962735.2 | XP_012818189.1 | 3 - 4 | 825 | 81 | ||
XM_012962736.2 | XP_012818190.1 | 3 - 4 | 822 | 81 | ||
XM_012962737.1 | XP_012818191.1 | 2 - 3 | 311 | 81 | ||
GenBank mRNA | BC161123.1 | - | 2 - 3 | 274 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |