Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlycd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126941.1 | NP_001120413.1 | 1 - 2 | 601 | 96 | |
GenBank mRNA | BC161140.1 | - | 1 - 2 | 601 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |