Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127973.2 | NP_001121445.2 | 2 - 3 | 249 | 81 | |
GenBank mRNA | BC166198.1 | - | 238 | 81 | ||
CX939571.1 | - | 2 - 3 | 249 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |