Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aste1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128015.1 | NP_001121487.1 | 3 - 4 | 1672 | 85 | |
GenBank mRNA | BC075288.1 | - | 1692 | 85 | ||
BC166275.1 | - | 3 - 4 | 1672 | 85 | ||
CR926284.2 | - | 3 - 4 | 1706 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |