Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c7orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128051.1 | NP_001121523.1 | 505 | 88 | ||
GenBank mRNA | BC166385.1 | - | 505 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |