Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128056.1 | NP_001121528.1 | 1 - 2 | 123 | 67 | |
XM_012960651.2 | XP_012816105.1 | 2 - 3 | 402 | 67 | ||
GenBank mRNA | BC166394.1 | - | 1 - 2 | 123 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |