Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130223.1 | NP_001123695.1 | 2 - 3 | 359 | 98 | |
XM_012962766.2 | XP_012818220.1 | 2 - 3 | 342 | 98 | ||
GenBank mRNA | BC167294.1 | - | 2 - 3 | 359 | 98 | |
BC167295.1 | - | 2 - 3 | 359 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |